Silver tetrafluoroborate 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Argentous tetrafluoroborate
197090100 197090500 197090010
ACRO197090100EA 3000 SEK
ACRO197090100 ACRO197090500 ACRO197090010
Silver tetrafluoroborate 99%
Silver tetrafluoroborate
Formel: AgBF₄
MW: 194.67 g/mol
Smältpunkt: 200
Storage Temperature: Omgivande
MDL Number: MFCD00003408
CAS nummer: 14104-20-2
EINECS: 237-956-5
UN: 3260
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Beige Crystalline powder
Assay ≥43.92 % (BF4)