1,8-Naphthalenedicarboxylic anhydride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Naphthalene-1,8-dicarboxylic anhydride, Benzo[de]isochromene-1,3-dione, 1,8-Naphthalic anhydride
A13519.0B A13519.18 A13519.30
A13519.0BEA 3425 SEK
A13519.0B A13519.18 A13519.30
1,8-Naphthalenedicarboxylic anhydride ≥97%
1,8-Naphthalenedicarboxylic anhydride
Purity: 97% CAS No.: 81-84-5 Molecular Formula: C12H6O3 Formula Weight: 198.18 Melting Point: 269-273° Beilstein Reference: 153190
Formel: C₁₂H₆O₃
MW: 198,18 g/mol
Kokpunkt: 425 °C (1013 hPa)
Smältpunkt: 269…273 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006925
CAS nummer: 81-84-5
EINECS: 201-380-2