Dansylklorid, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride
M-Clarity™ quality level = MQ100
1.03094.0001 1.03094.0010
1.03094.0001EA 713 SEK
1.03094.0001 1.03094.0010
Dansylklorid, Sigma-Aldrich®
Dansylklorid
Formel: C₁₂H₁₂ClNO₂S
MW: 269,75 g/mol
Smältpunkt: 69…73 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00003985
CAS nummer: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8,II
Merck Index: 13,02840