Dansylklorid 96%
Leverantör: Thermo Scientific
Fara
Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride
A13828.14 A13828.03 A13828.06
A13828.14EA 10350 SEK
A13828.14 A13828.03 A13828.06
Dansylklorid 96%
Dansylklorid
Purity: 96+% CAS No.: 605-65-2 Molecular Formula: C12H12ClNO2S Formula Weight: 269.75 Storage Temperature: 2°C to 8°C Odour: Odourless Beilstein Reference: 2217205 Merck Reference: 14,2814 UN No.: 3261 MDL No.: MFCD00003985
Formel: C₁₂H₁₂ClNO₂S
MW: 269,75 g/mol
Smältpunkt: 69…75 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00003985
CAS nummer: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8,II
Merck Index: 14,2814