6-Hydroxy-2-naphthoic acid ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 6-hydroxynaphthalene-2-carboxylic acid
B21791.06 B21791.14
B21791.06EA 1575 SEK
B21791.06 B21791.14
6-Hydroxy-2-naphthoic acid ≥99%
6-Hydroxy-2-naphthoic acid
Purity: 99% CAS No.: 16712-64-4 Molecular Formula: C11H8O3 Formula Weight: 188.18 Melting Point: 248-250° Beilstein Reference: 2692084
Formel: C₁₁H₈O₃
MW: 188,18 g/mol
Smältpunkt: 248…250 °C
Storage Temperature: Omgivande
MDL Number: MFCD00060070
CAS nummer: 16712-64-4
EINECS: 240-759-7