6-Bromo-2-naphthol 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene
107170250 107171000
ACRO107170250EA 1100 SEK
ACRO107170250 ACRO107171000
6-Bromo-2-naphthol 97%
6-Bromo-2-naphthol
Formel: C₁₀H₇BrO
MW: 223,07 g/mol
Kokpunkt: 200 - 205 °C (20 -rr)
Smältpunkt: 125…130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004081
CAS nummer: 15231-91-1
EINECS: 239-279-0
Order Now
Specifikation testresultat
Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 125 to 130 °C
GC ≥96.0 %
Color of solution ≤100 APHA (10% aq. soln.)