Dicerium trisulphide ≥99.9%
Leverantör: Thermo Scientific
Varning
Synonyms: Cerium (III) sulphide, Cerous sulphide
41856.06 41856.14
41856.06EA 2375 SEK
41856.06 41856.14
Dicerium trisulphide ≥99.9%
Dicerium trisulphide
Purity: 99.9% CAS No.: 12014-93-6 Molecular Formula: Ce2S3 Formula Weight: 376.43 Physical Form: -325 Mesh Powder Density: 5.02 Melting Point: 2100° UN No.: 3134 MDL No.: MFCD00016007
Formel: Ce₂S₃
MW: 376,43 g/mol
Smältpunkt: 2100 °C
Densitet: 5,02
Storage Temperature: Omgivande
MDL Number: MFCD00016007
CAS nummer: 12014-93-6
EINECS: 234-603-7
UN: 3134
ADR: 4.3,III