Praseodymium(III) nitrate hexahydrate 99,99 REO basis, REacton®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Praseodymium trinitrate hexahydrate
11240.14 11240.22
11240.14EA 4125 SEK
11240.14 11240.22
Praseodymium(III) nitrate hexahydrate 99,99 REO basis, REacton®
Praseodymium(III) nitrate hexahydrate
Purity: 99.99% CAS No.: 15878-77-0 Molecular Formula: Pr(NO3)3.6H2O Formula Weight: 435.01 (326.92anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic UN No.: 1477 MDL No.: MFCD00149826
Formel: N₃O₉Pr·6H₂O
MW: 435.01
Smältpunkt: 56
Densitet: 2.233 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149826
CAS nummer: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1 III