Molybdensyra ≥85% (MoO₃ basis) ACS, Fluka™
Leverantör: Honeywell Chemicals
Varning
232084-100G 232084-500G
232084-100GEA 1075 SEK
232084-100G 232084-500G
Molybdensyra ≥85% (MoO₃ basis) ACS, Fluka™
Molybdensyra
Formel: H₂MoO₄
Kokpunkt: ~ 300 °C (1013 hPa)
Smältpunkt: 300 °C
Densitet: 3,1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00036287
CAS nummer: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262
Order Now
Specifikation testresultat
Assay Min. 85.0 %
Insoluble in Ammonia Solution Max. 0.01 %
Heavy metals (as Pb) Max. 0.003 %
Phosphate, Arsenate, Silicate (as SiO₂) Max. 0.001 %
Chloride (Cl) Max. 0.002 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.2 %