Triphenylphosphine oxide

Leverantör: Thermo Scientific

BTB08974EE
MAYBBTB08974EEEA 0 SEK
MAYBBTB08974EE
Triphenylphosphine oxide
Triphenylphosphine oxide
Formel: C₁₈H₁₅OP
MW: 278,29 g/mol
Kokpunkt: ∼360 °C (1013 hPa)
Smältpunkt: 155…158 °C
Densitet: 1,2 g/cm³ (20 °C)
Flampunkt: 180 °C
MDL Number: MFCD00002080
CAS nummer: 791-28-6
EINECS: 212-338-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR