Tetrabutylammonium hexafluorophosphate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate
A17196.22 A17196.14 A17196.36
A17196.22EA 4975 SEK
A17196.22 A17196.14 A17196.36
Tetrabutylammonium hexafluorophosphate ≥98%
Tetrabutylammonium hexafluorophosphate
Purity: 98% CAS No.: 3109-63-5 Molecular Formula: C16H36F6NP Formula Weight: 387.44 Melting Point: 242-248° MDL No.: MFCD00011748
Formel: C₁₆H₃₆F₆NP
MW: 387,43 g/mol
Smältpunkt: 242…248 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011748
CAS nummer: 3109-63-5
EINECS: 221-472-6
UN: 3263
ADR: 8,III