Tetrahexylammonium iodide ≥99%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
Tetra-n-hexylammonium iodide
A17589.06
A17589.14
A17589.06EA
845
SEK
A17589.06
A17589.14
Tetrahexylammonium iodide ≥99%
Tetrahexylammonium iodide
Purity: 99%
CAS No.: 2138-24-1
Molecular Formula: [CH3(CH2)5]4NI
Formula Weight: 481.59
Melting Point: 99-105°
MDL No.: MFCD00041981
Formel:
C₂₄H₅₂IN MW: 481,59 g/mol Smältpunkt: 99…105 °C Densitet: 1,0922 g/cm³ (25 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00041981 CAS nummer: 2138-24-1 EINECS: 218-382-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...