Tetrabutylammonium perchlorate 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Tetra-n-butylammonium perchlorate
420140250 420141000 420145000
ACRO420140250EA 1225 SEK
ACRO420140250 ACRO420141000 ACRO420145000
Tetrabutylammonium perchlorate 98%
Tetrabutylammonium perchlorate
Formel: C₁₆H₃₆ClNO₄
MW: 341,92 g/mol
Smältpunkt: 210 °C
Densitet: 1,07 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00038722
CAS nummer: 1923-70-2
EINECS: 217-655-5
UN: 1479
ADR: 5.1,II
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration ≥97.5 %