Dodecyltrimethylammonium chloride ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC
A17924.06 A17924.14 A17924.22
A17924.06EA 620 SEK
A17924.06 A17924.14 A17924.22
Dodecyltrimethylammonium chloride ≥97%
Dodecyltrimethylammonium chloride
Purity: 97% CAS No.: 112-00-5 Molecular Formula: C15H34ClN Formula Weight: 263.90 Odour: Odourless Melting Point: ca 237° dec. Beilstein Reference: 3915951 UN No.: 3077 MDL No.: MFCD00041974
Formel: C₁₅H₃₄ClN
MW: 263,89 g/mol
Smältpunkt: ∼ 237 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00041974
CAS nummer: 112-00-5
EINECS: 203-927-0
ADR: 9,III