Boron carbide
Leverantör: Thermo Scientific
Varning
40504.18 40504.30
40504.18EA 1250 SEK
40504.18 40504.30
Boron carbide
Boron carbide
CAS No.: 12069-32-8 Molecular Formula: B4C Formula Weight: 55.26 Physical Form: 1-7 Micron Powder Density: 2.52 Odour: Odourless Merck Reference: 14,1344 MDL No.: MFCD00011520
Formel: CB₄
MW: 55,25 g/mol
Kokpunkt: ∼3500 °C (1013 hPa)
Smältpunkt: 2350 °C
Densitet: 2,52
Storage Temperature: Omgivande
MDL Number: MFCD00011520
CAS nummer: 12069-32-8
EINECS: 235-111-5
Merck Index: 14,1344