Barium titanate 99 Metaller basis, surface_area_high
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
41633.14 41633.22
41633.14EA 1975 SEK
41633.14 41633.22
Barium titanate 99 Metaller basis, surface_area_high
Barium titanate
Purity: 99+% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: 0.05-0.07 Micron APS Powder, S.A. 10-30m2/g Density: 5.85 Appearance: White to grey Odour: Odourless Melting Point: 1620° Merck Reference: 14,1000 MDL No.: MFCD00003447
Formel: BaO₃Ti
MW: 233.21 g/mol
Smältpunkt: 1620 °C
Densitet: 5.85
Storage Temperature: Omgivande
MDL Number: MFCD00003447
CAS nummer: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1 III
Merck Index: 14,1000