Kaliumvätedifluorid 99+%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Kaliumbifluorid, Kaliumvätefluorid
219230025 219230250 219235000
ACRO219230025EA 2575 SEK
ACRO219230025 ACRO219230250 ACRO219235000
Kaliumvätedifluorid 99+%, extra pure
Kaliumvätedifluorid
Formel: HF₂K
MW: 78,1 g/mol
Smältpunkt: 239 °C
Densitet: 2,37 g/cm³ (20 °C)
MDL Number: MFCD00011403
CAS nummer: 7789-29-9
EINECS: 232-156-2
UN: 1811
ADR: 8,II
Merck Index: 12,07771
Order Now
Specifikation testresultat
Appearance White to light grey Crystalline powder or crystals
Titration with NaOH ≥99.0 %
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm