1,2-Epoxycyclohexane 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Epoxycyclohexane, Cyclohexene oxide
111211000 111215000
ACRO111211000EA 410 SEK
ACRO111211000 ACRO111215000
1,2-Epoxycyclohexane 98%
1,2-Epoxycyclohexane
Formel: C₆H₁₀O
MW: 98,14 g/mol
Kokpunkt: 129 °C (1013 hPa)
Smältpunkt: –40 °C
Densitet: 0,97 g/cm³
Flampunkt: 27 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005162
CAS nummer: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %
BSC Certification Passes