Tetraklor-p-benzokinon ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Tetrachloroquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 4-Chloranil, p-Chloranil, 2,3,5,6-Tetrachlorobenzo-1,4-quinone, Tetrachloro-1,4-benzoquinone
A13495.14 A13495.22 A13495.36
A13495.14EA 795 SEK
A13495.14 A13495.22 A13495.36
Tetraklor-p-benzokinon ≥97%
Tetraklor-p-benzokinon
Purity: 97% CAS No.: 118-75-2 Molecular Formula: C6Cl4O2 Formula Weight: 245.88 Melting Point: ca 290° subl. Beilstein Reference: 393006 Merck Reference: 14,2078 UN No.: 3077 MDL No.: MFCD00001594
Formel: C₆Cl₄O₂
MW: 245,88 g/mol
Kokpunkt: 270 °C (1013 hPa)
Smältpunkt: ∼ 290 °C (subl.)
Densitet: 1,970
Flampunkt: >100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001594
CAS nummer: 118-75-2
EINECS: 204-274-4
ADR: 9,III
Merck Index: 14,2078