Oxybenzone ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)(phenyl)methanone
A17662.0B A17662.18 A17662.30
A17662.0BEA 6800 SEK
A17662.0B A17662.18 A17662.30
Oxybenzone ≥98%
Oxybenzone
Purity: 98+% CAS No.: 131-57-7 Molecular Formula: C14H12O3 Formula Weight: 228.25 Flash Point: 216°(420°F) Melting Point: 62-65° Boiling Point: ca 150°/5mm Beilstein Reference: 1913145 Merck Reference: 14,6954 MDL No.: MFCD00008387
Formel: C₁₄H₁₂O₃
MW: 228.25 g/mol
Kokpunkt: ∼150 °C (5 mmHg)
Smältpunkt: 62…65 °C
Densitet: 1.300
Flampunkt: 216 °C (420 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008387
CAS nummer: 131-57-7
EINECS: 205-031-5
Merck Index: 12,07088