Kloramin T (natriumsalt) trihydrat, white crystalline powder
Leverantör: MP Biomedicals
Fara
Synonyms: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
15011591 0215011583 0215011591
ICNA15011591EA 945 SEK
ICNA15011591 ICNA0215011583 ICNA0215011591
Kloramin T (natriumsalt) trihydrat, white crystalline powder
Kloramin T (natriumsalt) trihydrat
Soluble in water (100 mg/ml); practically insoluble in benzene, chloroform or ether.
  • Presentation : White Crystalline Powder
  • pH : 6 - 9(1% aq soln)
  • Detection Method : FTIR
  • Solubility :
Storage: protect from light, under nitrogen.
Formel: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281.69 g/mol
Smältpunkt: 167...170 °C (dec., lit.)
Densitet: 540...680 kg/m³ (bulk density)
Flampunkt: 192 °C (closed cup)
Storage Temperature: Kylskåp
MDL Number: MFCD00149066
CAS nummer: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8 III
Merck Index: 13,02085
Order Now
Specifikation testresultat
Identity Test Passes
FTIR Conforms to Standard
pH (5% aq soln) 8 - 10