4-Methoxybenzyl isothiocyanate 94%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-MBITC, 4-Methoxybenzylisothiocyanate
L12342.06 L12342.03
L12342.06EA 3275 SEK
L12342.06 L12342.03
4-Methoxybenzyl isothiocyanate 94%
4-Methoxybenzyl isothiocyanate
Purity: 94% CAS No.: 3694-57-3 Molecular Formula: C9H9NOS Formula Weight: 179.24 Refractive Index: 1.5975 Odour: Irritating Boiling Point: 132-133°/0.7mm Beilstein Reference: 2089549 UN No.: 2922 MDL No.: MFCD00041117
Formel: C₉H₉NOS
MW: 179,24 g/mol
Kokpunkt: 132…133 °C (0,7 mmHg)
Densitet: 1,150
Storage Temperature: Omgivande
MDL Number: MFCD00041117
CAS nummer: 3694-57-3
ADR: 8,III