Benzeneselenenyl chloride 98

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Phenylselenium chloride, Phenylselenenyl chloride, Chloroselenobenzene, Phenylselenyl chloride

A12751.06 A12751.14
A12751.06EA 1900 SEK
A12751.06 A12751.14
Benzeneselenenyl chloride 98
Benzeneselenenyl chloride

Purity: 98% CAS No.: 5707-04-0 Molecular Formula: C6H5ClSe Formula Weight: 191.52 Odour: Stench Melting Point: 60-65° Boiling Point: 120°/20mm Beilstein Reference: 1237091 UN No.: 2928 MDL No.: MFCD00000478

Formel: (C₆H₅)SeCl
MW: 191.52 g/mol
Kokpunkt: 120 °C (20 mmHg)
Smältpunkt: 60…65 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000478
CAS nummer: 5707-04-0
EINECS: 227-196-2
UN: 2928
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR