Dimethyl isophthalate 99%
Leverantör: Thermo Scientific
Synonyms: Isophthalic acid dimethyl ester
186510025 186510250 186515000
ACRO186510025EA 2300 SEK
ACRO186510025 ACRO186510250 ACRO186515000
Dimethyl isophthalate 99%
Dimethyl isophthalate
Formel: C₁₀H₁₀O₄
MW: 194,19 g/mol
Kokpunkt: 124 °C (16hPa)
Smältpunkt: >66 °C
Densitet: 1,186 g/cm³ (25 °C)
Flampunkt: 138 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008433
CAS nummer: 1459-93-4
EINECS: 215-951-9
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 60 to 70 °C
GC ≥98.5 %
Solubility (2 % in DMSO) Clear solution
HPLC ≥97.5 %