Gallussyra, propylester ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Propyl-3,4,5-trihydroxibenzoat
A10877.06 A10877.0E A10877.22 A10877.36
A10877.06EA 750 SEK
A10877.06 A10877.0E A10877.22 A10877.36
Gallussyra, propylester ≥98%
Gallussyra, propylester
Purity: 98% CAS No.: 121-79-9 Molecular Formula: C10H12O5 Formula Weight: 212.20 Physical Form: Powder Appearance: White Melting Point: 146-150° Beilstein Reference: 1877976 Merck Reference: 14,7859 MDL No.: MFCD00002196
Formel: C₁₀H₁₂O₅
MW: 212.2 g/mol
Kokpunkt: 449
Smältpunkt: 146…150 °C
Densitet: 1.21 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002196
CAS nummer: 121-79-9
EINECS: 204-498-2
Merck Index: 12,08044