Oktyl-3,4,5-trihydroxibenzoat ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Octyl 3,4,5-trihydroxybenzoate, Gallic acid octyl ester, Octyl gallate, n-Octyl gallate
L06081.14 L06081.22
L06081.14EA 1200 SEK
L06081.14 L06081.22
Oktyl-3,4,5-trihydroxibenzoat ≥98%
Oktyl-3,4,5-trihydroxibenzoat
Purity: 98+% CAS No.: 1034-01-1 Molecular Formula: C15H22O5 Formula Weight: 282.34 Melting Point: 96-102° Beilstein Reference: 2132305
Formel: C₁₅H₂₂O₅
MW: 282,34 g/mol
Smältpunkt: 96…102 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002197
CAS nummer: 1034-01-1
EINECS: 213-853-0