Methyl-3,4,5-trihydroxybenzoate 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl gallate, Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate
243460050 243460500 243462500
ACRO243460050EA 750 SEK
ACRO243460050 ACRO243460500 ACRO243462500
Methyl-3,4,5-trihydroxybenzoate 99%
Methyl-3,4,5-trihydroxybenzoate
Formel: C₈H₈O₅
MW: 184,15 g/mol
Kokpunkt: 450 °C (1013 hPa)
Smältpunkt: 199…203 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002194
CAS nummer: 99-24-1
EINECS: 202-741-7
Merck Index: 13,04366
Order Now
Specifikation testresultat
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 199 to 203 °C
HPLC ≥98.5 %