Isophthaloyl dichloride 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Isophthaloyl chloride
122661000 122665000
ACRO122661000EA 520 SEK
ACRO122661000 ACRO122665000
Isophthaloyl dichloride 98%
Isophthaloyl dichloride
Formel: C₈H₄Cl₂O₂
MW: 203.02 g/mol
Kokpunkt: 276 °C (760 mmHg)
Smältpunkt: 43…45 °C
Flampunkt: 180 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000678
CAS nummer: 99-63-8
EINECS: 202-774-7
UN: 2928
ADR: 6.1 I
Order Now
Specifikation testresultat
Appearance White Crystalline mass
Infrared spectrum Conforms
Melting point 43 to 45 °C
Titration RCOCl ≥97.5 %
NMR Consistent with structure
Specific optical rotation +30° to +33° (20 °C, 589 nm) (C= neat)