4-Chloro-3-nitrophenyl isothiocyanate

Leverantör: Thermo Scientific

Synonyms: 1-Chloro-4-isothiocyanato-3-nitrobenzene

SB01784EA
MAYBSB01784EAEA 0 SEK
MAYBSB01784EA
4-Chloro-3-nitrophenyl isothiocyanate
4-Chloro-3-nitrophenyl isothiocyanate
Formel: C₇H₃ClN₂O₂S
Smältpunkt: 53…54 °C
MDL Number: MFCD00041236
CAS nummer: 127142-66-9
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR