Zaltoprofen ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-(10,11-Dihydro-10-oxodibenzo[b,f]thiepin-2-yl)propionic Acid, 2-(10-Oxo-10,11-dihydrodibenzo[b,f]thiepin-2-yl)propanoic acid
Z0021-1G
Z0021-5G
TCIAZ0021-1GEA
750
SEK
TCIAZ0021-1G
TCIAZ0021-5G
Zaltoprofen ≥98.0% (by HPLC, titration analysis)
Zaltoprofen
Formel:
C₁₇H₁₄O₃S MW: 298,36 g/mol |
MDL Number:
MFCD00864323 CAS nummer: 74711-43-6 EINECS: 277-973-5 Merck Index: 14,10112 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...