Ytterbium(III)oxid ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Diytterbium trioxide, Ytterbium oxide
Y0008-5G
Y0008-25G
TCIAY0008-5GEA
295
SEK
TCIAY0008-5G
TCIAY0008-25G
Ytterbium(III)oxid ≥98.0%
Ytterbium(III) oxide
Formel:
O₃Yb₂ MW: 394,08 g/mol |
MDL Number:
MFCD00011290 CAS nummer: 1314-37-0 EINECS: 215-234-0 Merck Index: 14,10105 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...