(S)-(-)-XylBINAP ≥97.0%

Leverantör: TCI EUROPE

Synonyms: (S)-2,2'-Bis(bis(3,5-dimethylphenyl)phosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl

X0071-200MG X0071-1G
TCIAX0071-200MGEA 430 SEK
TCIAX0071-200MG TCIAX0071-1G
(S)-(-)-XylBINAP ≥97.0%
(S)-(-)-XylBINAP
Formel: C₅₂H₄₈P₂
MW: 734,90 g/mol
Smältpunkt: 255°C
MDL Number: MFCD01630821
CAS nummer: 135139-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0b1fc5ffe0e1257f25ea0522d25f3cb7f78a7df&vid=d0b1fc5ffe0e1257f25ea0522d25f3cb7f78a7df&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Optical purity (LC) min. 99.0 ee%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR