Xylosucrose ≥98.0%

Leverantör: TCI EUROPE

Synonyms: beta-D-Fructofuranosyl alpha-D-Xylopyranoside

X0065-1G X0065-100MG
TCIAX0065-1GEA 4350 SEK
TCIAX0065-1G TCIAX0065-100MG
Xylosucrose ≥98.0%
Xylosucrose
Formel: C₁₁H₂₀O₁₀
MW: 312,27 g/mol
CAS nummer: 512-66-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 112.0 to 116.0 deg-C
Specific rotation [a]20/D +53.0 to +57.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR