p-Xylylene dithiocyanate ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: alpha,alpha'-Dithiocyanato-p-xylene

X0060-25G X0060-5G
TCIAX0060-25GEA 4750 SEK
TCIAX0060-25G TCIAX0060-5G
p-Xylylene dithiocyanate ≥98.0%
p-Xylylene dithiocyanate
Formel: C₁₀H₈N₂S₂
MW: 220,31 g/mol
CAS nummer: 62855-80-5
UN: 2811
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR