m-Xylylene diisocyanate ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1,3-Bis(isocyanatomethyl)benzene, Isocyanic Acid 1,3-Phenylenedimethylene Ester, m-Xylene Diisocyanate, M-xylylenediisocyanate

X0022-25G X0022-500G X0022-100G
TCIAX0022-25GEA 230 SEK
TCIAX0022-25G TCIAX0022-500G TCIAX0022-100G
m-Xylylene diisocyanate ≥98.0%
m-Xylylene diisocyanate
Formel: C₁₀H₈N₂O₂
MW: 188,19 g/mol
Kokpunkt: 126°C
Flampunkt: 174°C
Storage Temperature: Kylskåp
MDL Number: MFCD00019917
CAS nummer: 3634-83-1
EINECS: 222-852-4
UN: 2206
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78c564535da4754a5db9a1b354075857e9dad5bd&vid=78c564535da4754a5db9a1b354075857e9dad5bd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR