Wogonin ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 5,7-Dihydroxy-8-methoxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxy-8-methoxyflavone

W0010-50MG W0010-250MG
TCIAW0010-50MGEA 1575 SEK
TCIAW0010-50MG TCIAW0010-250MG
Wogonin ≥98.0%
Wogonin
Formel: C₁₆H₁₂O₅
MW: 284,3 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00017736
CAS nummer: 632-85-9

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 205.0 to 209.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR