Warfarin sodium 98,0 HPLC_ANALYSMETOD (contains Isopropyl Alcohol)
Leverantör: TCI
|
Fara
|
Synonyms:
3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin
W0005-25G
W0005-5G
TCIAW0005-25GEA
1825
SEK
TCIAW0005-25G
TCIAW0005-5G
Warfarin sodium 98,0 HPLC_ANALYSMETOD (contains Isopropyl Alcohol)
Warfarin sodium
Formel:
C₁₉H₁₆NaO₄ MW: 331.33 g/mol |
MDL Number:
MFCD00083223 CAS nummer: 129-06-6 EINECS: 204-929-4 UN: 1544 ADR: 6.1 II Merck Index: 14,10038 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 91.0 % |
2-Propanol | 7.0 to 9.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...