VGX-1027 ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
VGX 1027, 2-(3-Phenyl-4,5-dihydroisoxazol-5-yl)acetic acid, VGX-1027, VGX1027, GIT 27, GIT-27, 2-(3-phenyl-4,5-dihydro-1,2-oxazol-5-yl)acetic acid, GIT27
V0160-10MG
V0160-50MG
TCIAV0160-10MGEA
1350
SEK
TCIAV0160-10MG
TCIAV0160-50MG
VGX-1027 ≥98.0%
VGX-1027
Formel:
C₁₁H₁₁NO₃ MW: 205,21 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD08696167 CAS nummer: 6501-72-0 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...