Verbascoside ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Acteoside, 2-(3,4-Dihydroxyphenyl)ethyl 3-O-(6-Deoxy-alpha-L-mannopyranosyl)-4-O-[(2E)-3-(3,4-dihydroxyphenyl)-2-propenoyl]-beta-D-glucopyranoside, (2R,3R,4R,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-5-hydroxy-2-(hydroxymethyl)-4-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl (E)-3-(3,4-dihydroxyphenyl)acrylate, Kusaginin

V0153-10MG V0153-50MG
TCIAV0153-10MGEA 430 SEK
TCIAV0153-10MG TCIAV0153-50MG
Verbascoside ≥97.0%
Verbascoside
Formel: C₂₉H₃₆O₁₅
MW: 624,59 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00221751
CAS nummer: 61276-17-3

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystaline
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D -87.0 to -97.0 deg (C=0.1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR