Vilazodone hydrochloride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 5-[4-[4-(5-Cyano-1H-indol-3-yl)butyl]piperazin-1-yl]benzofuran-2-carboxamide hydrochloride

V0149-50MG V0149-200MG
TCIAV0149-50MGEA 645 SEK
TCIAV0149-50MG TCIAV0149-200MG
Vilazodone hydrochloride ≥98.0%
Vilazodone hydrochloride
Formel: C₂₆H₂₇N₅O₂·HCl
MW: 477,99 g/mol
Smältpunkt: 279°C
Storage Temperature: Frys
MDL Number: MFCD00940014
CAS nummer: 163521-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d119112a904332fa355ed172a29b09da5f6691ab&vid=d119112a904332fa355ed172a29b09da5f6691ab&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 14.00 to 15.40 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR