2,2'-(2-Vinylanthracene-9,10-diylidene)dimalononitrile ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
9,10-Bis(dicyanomethylene)-2-vinylanthracene
V0137-1G
V0137-5G
TCIAV0137-1GEA
1125
SEK
TCIAV0137-1G
TCIAV0137-5G
2,2'-(2-Vinylanthracene-9,10-diylidene)dimalononitrile ≥98.0%
2,2'-(2-Vinylanthracene-9,10-diylidene)dimalononitrile
Formel:
C₂₂H₁₀N₄ |
CAS nummer:
1612793-07-3 UN: 3439 ADR: 6.1,III |
|
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 16.00 to 18.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...