2,2'-(2-Vinylanthracene-9,10-diylidene)bis(1,3-dithiole) ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 9,10-Bis(1,3-dithiol-2-ylidene)-2-vinylanthracene

V0136-100MG V0136-500MG
TCIAV0136-100MGEA 1500 SEK
TCIAV0136-100MG TCIAV0136-500MG
2,2'-(2-Vinylanthracene-9,10-diylidene)bis(1,3-dithiole) ≥98.0%
2,2'-(2-Vinylanthracene-9,10-diylidene)bis(1,3-dithiole)
Formel: C₂₂H₁₄S₄
MW: 406,59 g/mol
CAS nummer: 736998-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98082c0650684d1246d3a440a7d53f65248e07a0&vid=98082c0650684d1246d3a440a7d53f65248e07a0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR