vBRIDP® ≥98.0%

Leverantör: TCI

Synonyms: Di-tert-butyl(1,1-diphenyl-1-propen-2-yl)phosphine

V0132-200MG V0132-1G
TCIAV0132-200MGEA 310 SEK
TCIAV0132-200MG TCIAV0132-1G
vBRIDP® ≥98.0%
vBRIDP®
Formel: C₂₃H₃₁P
Smältpunkt: 133°C
MDL Number: MFCD16621412
CAS nummer: 384842-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74ad6655fa316ae072fc5cf9ec7bcaf6e718ca48&vid=74ad6655fa316ae072fc5cf9ec7bcaf6e718ca48&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR