9-Vinylanthracene ≥98.0%

Leverantör: TCI EUROPE

V0130-1G V0130-5G
TCIAV0130-1GEA 575 SEK
TCIAV0130-1G TCIAV0130-5G
9-Vinylanthracene ≥98.0%
9-Vinylanthracene
Formel: C₁₆H₁₂
MW: 204,27 g/mol
Kokpunkt: 145°C
Storage Temperature: Kylskåp
MDL Number: MFCD00003576
CAS nummer: 2444-68-0
EINECS: 219-486-2

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR