Valsartan ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: (S)-2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid, (S)-2-(N-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid

V0112-1G V0112-5G
TCIAV0112-1GEA 495 SEK
TCIAV0112-1G TCIAV0112-5G
Valsartan ≥98.0% (by HPLC, titration analysis)
Valsartan
Formel: C₂₄H₂₉N₅O₃
MW: 435,52 g/mol
Smältpunkt: 117°C
MDL Number: MFCD00865840
CAS nummer: 137862-53-4
Merck Index: 14,09916

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -63.0 to -67.0 deg (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR