3-Vinylpyridine ≥96.0% (by GC, titration analysis) stabilised

Leverantör: TCI
Fara


Stabilization: med 4-tert-Butylpyrokatekol

V0099-1G V0099-5G
TCIAV0099-1GEA 2300 SEK
TCIAV0099-1G TCIAV0099-5G
3-Vinylpyridine ≥96.0% (by GC, titration analysis) stabilised
3-Vinylpyridine
Formel: C₇H₇N
Kokpunkt: 82°C
Storage Temperature: Frys
MDL Number: MFCD02178019
CAS nummer: 1121-55-7
UN: 3073
ADR: 6.1,II
Merck Index: 14,03730

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR