Vinylene carbonate ≥98.0% stabilised
Leverantör: TCI
Stabilization:
Stabilised med BHT/Ionol
V0015-25G
V0015-5G
TCIAV0015-25GEA
1725
SEK
TCIAV0015-25G
TCIAV0015-5G
Vinylene carbonate ≥98.0% stabilised
Vinylene carbonate
Formel:
C₃H₂O₃ Kokpunkt: 60°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00005380 CAS nummer: 872-36-6 EINECS: 212-825-5 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...