sulphur Dioxide 1-Methylpyrrolidine adduct ≥96.0%
Leverantör: TCI
|
Fara
|
U0113-25G
U0113-5G
TCIAU0113-25GEA
2650
SEK
TCIAU0113-25G
TCIAU0113-5G
sulphur Dioxide 1-Methylpyrrolidine adduct ≥96.0%
sulphur Dioxide 1-Methylpyrrolidine
Formel:
C₅H₁₁NO₂S Storage Temperature: Frys |
CAS nummer:
2098851-56-8 UN: 1993 ADR: 3,II |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...