Sulconazole nitrate ≥98.0%

Leverantör: TCI
Varning

U0112-250MG U0112-50MG
TCIAU0112-250MGEA 3400 SEK
TCIAU0112-250MG TCIAU0112-50MG
Sulconazole nitrate ≥98.0%
Sulconazole nitrate
Formel: C₁₈H₁₅Cl₃N₂S·HNO₃
MDL Number: MFCD00058163
CAS nummer: 61318-91-0
Merck Index: 14,08893

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.60 to 9.60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR