SU 1498 ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
SU1498, (E)-2-Cyano-3-(4-hydroxy-3,5-diisopropylphenyl)-N-(3-phenylpropyl)acrylamide
U0103-25MG
U0103-5MG
TCIAU0103-25MGEA
4325
SEK
TCIAU0103-25MG
TCIAU0103-5MG
SU 1498 ≥98.0%
SU 1498
Formel:
C₂₅H₃₀N₂O₂ MW: 390,52 g/mol Storage Temperature: Frys |
MDL Number:
MFCD03453026 CAS nummer: 168835-82-3 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 124.0 to 128.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...